2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-acetamidine hydrochloride

Supplier: Apollo Scientific

PC446066-1G PC446066-5G
APOSPC446066-1GEA 308 EUR
APOSPC446066-1G APOSPC446066-5G
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-acetamidine hydrochloride
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-acetamidine hydrochloride
Formula: C₈H₇ClF₃N₃·ClH
MW: 274,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981494
CAS Number: 1332495-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR