3-(3-Chloro-2-oxo-5-(trifluoromethyl)-2H-pyridin-1-yl)propionitrile 95+%
Supplier: Apollo Scientific
PC446084-1G
PC446084-5G
APOSPC446084-1GEA
154
EUR
APOSPC446084-1G
APOSPC446084-5G
3-(3-Chloro-2-oxo-5-(trifluoromethyl)-2H-pyridin-1-yl)propionitrile 95+%
3-(3-Chloro-2-oxo-5-(trifluoromethyl)-2H-pyridin-1-yl)propionitrile
Formula:
C₉H₆ClF₃N₂O MW: 250,605 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568269 CAS Number: 1053658-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...