(Z)-3-Amino-3-(2-fluorophenyl)acrylonitrile 95+%

Supplier: Apollo Scientific

PC446098-1G PC446098-5G
APOSPC446098-1GEA 201 EUR
APOSPC446098-1G APOSPC446098-5G
(Z)-3-Amino-3-(2-fluorophenyl)acrylonitrile 95+%
(Z)-3-Amino-3-(2-fluorophenyl)acrylonitrile
Formula: C₉H₇FN₂
MW: 162,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12131141
CAS Number: 71682-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR