2-Chloro-6-(trifluoromethyl)pyridine-3-sulphonyl chloride 95+%
Supplier: Apollo Scientific
PC446101-1G
PC446101-500MG
APOSPC446101-1GEA
528
EUR
APOSPC446101-1G
APOSPC446101-500MG
2-Chloro-6-(trifluoromethyl)pyridine-3-sulphonyl chloride 95+%
2-Chloro-6-(trifluoromethyl)pyridine-3-sulphonyl chloride
Formula:
C₆H₂Cl₂F₃NO₂S MW: 280,052 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15142816 CAS Number: 1208081-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...