(2-Fluorophenoxy)acetonitrile 95+%
Supplier: Apollo Scientific
Synonyms:
2-Fluorophenoxyacetonitrile
PC446174-1G
PC446174-25G
PC446174-5G
APOSPC446174-1GEA
118
EUR
APOSPC446174-1G
APOSPC446174-25G
APOSPC446174-5G
(2-Fluorophenoxy)acetonitrile 95+%
(2-Fluorophenoxy)acetonitrile
Formula:
C₈H₆FNO MW: 151,138 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09941337 CAS Number: 137988-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...