α-Methyl-2-(trifluoromethyl)benzylamine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(1-Aminoethyl)benzotrifluoride, 1-(2-Trifluoromethylphenyl)ethylamine, 1-[2-(Trifluoromethyl)phenyl]ethylamine

PC4462-1G PC4462-5G PC4462-250MG
APOSPC4462-1GEA 125 EUR
APOSPC4462-1G APOSPC4462-5G APOSPC4462-250MG
α-Methyl-2-(trifluoromethyl)benzylamine 97%
α-Methyl-2-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃N
MW: 189,178 g/mol
Boiling Pt: 68…72 °C (7,8 mm Hg)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD04038876
CAS Number: 273384-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR