2-Fluoro-3-hydroxybenzaldehyde 98%

Supplier: Apollo Scientific

PC448018-1G PC448018-5G
APOSPC448018-1GEA 109 EUR
APOSPC448018-1G APOSPC448018-5G
2-Fluoro-3-hydroxybenzaldehyde 98%
2-Fluoro-3-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140,112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110191
CAS Number: 103438-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR