2-Amino-N-(2,2,2-trifluoroethyl)acetamide

Supplier: Apollo Scientific
Warning

PC450007-1G PC450007-250MG PC450007-5G
APOSPC450007-1GEA 68.4 EUR
APOSPC450007-1G APOSPC450007-250MG APOSPC450007-5G
2-Amino-N-(2,2,2-trifluoroethyl)acetamide
2-Amino-N-(2,2,2-trifluoroethyl)acetamide
Formula: C₄H₇F₃N₂O
MW: 156,106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09944516
CAS Number: 359821-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR