1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorooctane pentyl ether
Supplier: Apollo Scientific
PC450094-1G
PC450094-5G
APOSPC450094-1GEA
80.3
EUR
APOSPC450094-1G
APOSPC450094-5G
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorooctane pentyl ether
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorooctane pentyl ether
Formula:
C₁₃H₁₅F₁₃O Storage Temperature: Ambient |
MDL Number:
MFCD16619395 CAS Number: 1193009-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...