2,2,3,3,4,4,5,5-Octafluoropentyl difluoromethylether 97%

Supplier: Apollo Scientific
Warning

PC450102-1G PC450102-250MG
APOSPC450102-1GEA 198 EUR
APOSPC450102-1G APOSPC450102-250MG
2,2,3,3,4,4,5,5-Octafluoropentyl difluoromethylether 97%
2,2,3,3,4,4,5,5-Octafluoropentyl difluoromethylether
Formula: C₆H₄F₁₀O
MW: 282,079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27918474
CAS Number: 742063-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR