2,2,3,3,4,4,4-Heptafluorobutyl difluoromethylether
Supplier: Apollo Scientific
PC450111-1G
PC450111-250MG
APOSPC450111-1GEA
227
EUR
APOSPC450111-1G
APOSPC450111-250MG
2,2,3,3,4,4,4-Heptafluorobutyl difluoromethylether
2,2,3,3,4,4,4-Heptafluorobutyl difluoromethylether
Formula:
C₅H₃F₉O MW: 250,062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27918473 CAS Number: 50428-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...