2,2-Difluoro-(2H)-1,4-benzothiazin-3(4H)-one
Supplier: Apollo Scientific
PC450120-1G
PC450120-250MG
APOSPC450120-1GEA
546
EUR
APOSPC450120-1G
APOSPC450120-250MG
2,2-Difluoro-(2H)-1,4-benzothiazin-3(4H)-one
2,2-Difluoro-(2H)-1,4-benzothiazin-3(4H)-one
Formula:
C₈H₅F₂NOS MW: 201,193 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12404008 CAS Number: 1216262-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...