1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene

Supplier: Apollo Scientific
Warning

PC450160-1G PC450160-5G
APOSPC450160-1GEA 102 EUR
APOSPC450160-1G APOSPC450160-5G
1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene
1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene
Formula: C₉H₆BrF₅O
MW: 305,039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054311
CAS Number: 1823420-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR