Sodium 2,2-difluorobutyrate

Supplier: Apollo Scientific

PC450208-1G PC450208-5G
APOSPC450208-1GEA 139 EUR
APOSPC450208-1G APOSPC450208-5G
Sodium 2,2-difluorobutyrate
Sodium 2,2-difluorobutyrate
Formula: C₄H₅F₂O₂·Na
MW: 146,068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486155
CAS Number: 1346521-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR