Bis(2,2,3,3-tetrafluoropropyl) sulphite

Supplier: Apollo Scientific

PC450225-1G PC450225-25G PC450225-5G
APOSPC450225-1GEA 65.5 EUR
APOSPC450225-1G APOSPC450225-25G APOSPC450225-5G
Bis(2,2,3,3-tetrafluoropropyl) sulphite
Bis(2,2,3,3-tetrafluoropropyl) sulphite
Formula: C₆H₆F₈O₃S
MW: 310,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25978122
CAS Number: 649-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR