Perfluorotetrahydro-2-furancarboxylic acid
Supplier: Apollo Scientific
|
Danger
|
PC450231-1G
PC450231-5G
APOSPC450231-1GEA
158
EUR
APOSPC450231-1G
APOSPC450231-5G
Perfluorotetrahydro-2-furancarboxylic acid
Perfluorotetrahydro-2-furancarboxylic acid
Formula:
C₅HF₇O₃ MW: 242,048 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28054325 CAS Number: 65578-62-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...