2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride
Supplier: Apollo Scientific
|
Danger
|
PC450255-1G
PC450255-25G
PC450255-5G
APOSPC450255-1GEA
36.4
EUR
APOSPC450255-1G
APOSPC450255-25G
APOSPC450255-5G
2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride
2,2,3,3,4,4,4-Heptafluorobutylamine hydrochloride
Formula:
C₄H₄F₇N·ClH Storage Temperature: Ambient |
MDL Number:
MFCD22124010 CAS Number: 2794-75-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...