Propyl 1,1,2,2-tetrafluoroethyl ether

Supplier: Apollo Scientific
Danger

PC450273-100G PC450273-25G
APOSPC450273-100GEA 234 EUR
APOSPC450273-100G APOSPC450273-25G
Propyl 1,1,2,2-tetrafluoroethyl ether
Propyl 1,1,2,2-tetrafluoroethyl ether
Formula: C₅H₈F₄O
MW: 160,11 g/mol
Boiling Pt: 74 °C
Density: 1,128 g/cm³ (25 °C)
Flash Pt: –4,5 °C
Storage Temperature: Ambient
MDL Number: MFCD16619128
CAS Number: 380-48-3
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR