4-Fluoroisophthalic acid
Supplier: Apollo Scientific
|
Warning
|
PC450277-1G
PC450277-25G
PC450277-5G
APOSPC450277-1GEA
116
EUR
APOSPC450277-1G
APOSPC450277-25G
APOSPC450277-5G
4-Fluoroisophthalic acid
4-Fluoroisophthalic acid
Formula:
C₈H₅FO₄ Melting Pt: 290…293 °C Storage Temperature: Ambient |
MDL Number:
MFCD06208805 CAS Number: 327-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...