Isopropyl 1,1,2,2-tetrafluoroethyl ether
Supplier: Apollo Scientific
PC450281-100G
PC450281-25G
PC450281-5G
APOSPC450281-100GEA
542
EUR
APOSPC450281-100G
APOSPC450281-25G
APOSPC450281-5G
Isopropyl 1,1,2,2-tetrafluoroethyl ether
Isopropyl 1,1,2,2-tetrafluoroethyl ether
Formula:
C₅H₈F₄O MW: 160,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16619130 CAS Number: 757-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...