Sodium 2,2-difluoropropionate
Supplier: Apollo Scientific
PC450283-1G
PC450283-5G
APOSPC450283-1GEA
69.8
EUR
APOSPC450283-1G
APOSPC450283-5G
Sodium 2,2-difluoropropionate
Sodium 2,2-difluoropropionate
Formula:
C₃H₃F₂O₂·Na MW: 132,041 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20489367 CAS Number: 1281943-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...