7-Fluoronaphthalen-1-ol 95%

Supplier: Apollo Scientific

PC45032-100MG PC45032-250MG
APOSPC45032-100MGEA 165 EUR
APOSPC45032-100MG APOSPC45032-250MG
7-Fluoronaphthalen-1-ol 95%
7-Fluoronaphthalen-1-ol
Formula: C₁₀H₇FO
MW: 162,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077540
CAS Number: 3132-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR