2,2-Difluoroethyl 1H-imidazole-1-carboxylate, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
PC450492-1G
PC450492-5G
APOSPC450492-1GEA
109
EUR
APOSPC450492-1G
APOSPC450492-5G
2,2-Difluoroethyl 1H-imidazole-1-carboxylate, Technical Grade
2,2-Difluoroethyl 1H-imidazole-1-carboxylate
Formula:
C₆H₆F₂N₂O₂ MW: 176,121 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28334494 CAS Number: 1980034-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...