(2-Chloro-1-ethoxy-2,2-difluoroethoxy)triethylsilane
Supplier: Apollo Scientific
PC450510-1G
PC450510-5G
APOSPC450510-1GEA
248
EUR
APOSPC450510-1G
APOSPC450510-5G
(2-Chloro-1-ethoxy-2,2-difluoroethoxy)triethylsilane
(2-Chloro-1-ethoxy-2,2-difluoroethoxy)triethylsilane
Formula:
C₁₀H₂₁ClF₂O₂Si MW: 274,808 g/mol Boiling Pt: 78…79 °C (6 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD30730297 CAS Number: 1922958-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...