(1-Ethoxy-2,2,3,3,3-pentafluoropropoxy) triethylsilane

Supplier: Apollo Scientific

PC450513-1G PC450513-5G
APOSPC450513-1GEA 248 EUR
APOSPC450513-1G APOSPC450513-5G
(1-Ethoxy-2,2,3,3,3-pentafluoropropoxy) triethylsilane
(1-Ethoxy-2,2,3,3,3-pentafluoropropoxy) triethylsilane
Formula: C₁₁H₂₁F₅O₂Si
MW: 308,361 g/mol
Boiling Pt: 86…87 °C (15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD30730300
CAS Number: 1922958-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR