1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane 97%
Supplier: Apollo Scientific
PC450526-250MG
PC450526-1G
APOSPC450526-250MGEA
219
EUR
APOSPC450526-250MG
APOSPC450526-1G
1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane 97%
1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane
Formula:
C₈H₁₉F₃O₂Si₂ MW: 260,401 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30730309 CAS Number: 2149590-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...