Trifluorovinyltrimethylsilane

Supplier: Apollo Scientific
Danger

PC450570-1G PC450570-250MG PC450570-5G
APOSPC450570-1GEA 1010 EUR
APOSPC450570-1G APOSPC450570-250MG APOSPC450570-5G
Trifluorovinyltrimethylsilane
Trifluorovinyltrimethylsilane
Formula: C₅H₉F₃Si
MW: 154.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30062985
CAS Number: 1427-33-4
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR