2-Iodo-1,1,1,4,4,4-hexafluorobutane 97%
Supplier: Apollo Scientific
|
Warning
|
PC4515-1G
PC4515-5G
APOSPC4515-1GEA
73
EUR
APOSPC4515-1G
APOSPC4515-5G
2-Iodo-1,1,1,4,4,4-hexafluorobutane 97%
2-Iodo-1,1,1,4,4,4-hexafluorobutane
Formula:
C₄H₃F₆I Boiling Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458098 CAS Number: 453-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...