3-[(1H,1H,9H-Perfluoronon-1-yl)oxy]-1,2-propenoxide 96%

Supplier: Apollo Scientific
Warning

Synonyms: 2-{[(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoronon-1-yl)oxy]methyl}oxirane

PC4535B-25G PC4535B-5G
APOSPC4535B-25GEA 720 EUR
APOSPC4535B-25G APOSPC4535B-5G
3-[(1H,1H,9H-Perfluoronon-1-yl)oxy]-1,2-propenoxide 96%
3-[(1H,1H,9H-Perfluoronon-1-yl)oxy]-1,2-propenoxide
Formula: C₁₂H₈F₁₆O₂
MW: 488,165 g/mol
Boiling Pt: 251 °C
Density: 1,682 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00236091
CAS Number: 125370-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR