6-Bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromo-2,2,3,3-tetrafluoro-2,3-dihydro-1,4-benzodioxine
PC4567-1G
PC4567-250MG
APOSPC4567-1GEA
783
EUR
APOSPC4567-1G
APOSPC4567-250MG
6-Bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-Bromo-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formula:
C₈H₃BrF₄O₂ MW: 287,006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236309 CAS Number: 141872-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...