6-Chloro-1H-perfluorohexane 97%

Supplier: Apollo Scientific

PC4677-1G PC4677-5G
APOSPC4677-1GEA 109 EUR
APOSPC4677-1G APOSPC4677-5G
6-Chloro-1H-perfluorohexane 97%
6-Chloro-1H-perfluorohexane
Formula: C₆HClF₁₂
MW: 336,506 g/mol
Boiling Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00153754
CAS Number: 307-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR