6-(2,2,2-Trifluoroethoxy)nicotinonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
PC4821-1G
PC4821-5G
APOSPC4821-1GEA
139
EUR
APOSPC4821-1G
APOSPC4821-5G
6-(2,2,2-Trifluoroethoxy)nicotinonitrile 98%
6-(2,2,2-Trifluoroethoxy)nicotinonitrile
Formula:
C₈H₅F₃N₂O MW: 202,133 g/mol Melting Pt: 47…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100036 CAS Number: 159981-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...