4',5'-Dichloro-2'-fluoroacetophenone 99%

Supplier: Apollo Scientific
Warning

PC48405-1G PC48405-5G
APOSPC48405-1GEA 71.2 EUR
APOSPC48405-1G APOSPC48405-5G
4',5'-Dichloro-2'-fluoroacetophenone 99%
4',5'-Dichloro-2'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
Melting Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD18632530
CAS Number: 1286734-89-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR