6-Bromo-3-chloro-2-fluorobenzeneboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromo-3-chloro-2-fluorophenylboronic acid

PC48528-1G PC48528-250MG PC48528-5G
APOSPC48528-1GEA 29 EUR
APOSPC48528-1G APOSPC48528-250MG APOSPC48528-5G
6-Bromo-3-chloro-2-fluorobenzeneboronic acid
6-Bromo-3-chloro-2-fluorobenzeneboronic acid
Formula: C₆H₄BBrClFO₂
MW: 253,261 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111906
CAS Number: 1451392-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR