7-Bromo-8-fluoroquinoline 95%

Supplier: Apollo Scientific
Warning

PC48659-1G PC48659-5G
APOSPC48659-1GEA 699 EUR
APOSPC48659-1G APOSPC48659-5G
7-Bromo-8-fluoroquinoline 95%
7-Bromo-8-fluoroquinoline
Formula: C₉H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD23163685
CAS Number: 1375107-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR