Methyl 3-amino-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Supplier: Apollo Scientific
Warning

PC48669-1G PC48669-250MG
APOSPC48669-1GEA 208 EUR
APOSPC48669-1G APOSPC48669-250MG
Methyl 3-amino-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Methyl 3-amino-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formula: C₁₁H₈F₃NO₂S
MW: 275,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032272
CAS Number: 35212-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR