3-[(Trifluoromethyl)sulphonyl]aniline 97%

Supplier: Apollo Scientific
Danger

Synonyms: 3-Aminophenyl trifluoromethyl sulphone

PC4871-1G PC4871-250MG PC4871-5G
APOSPC4871-1GEA 132 EUR
APOSPC4871-1G APOSPC4871-250MG APOSPC4871-5G
3-[(Trifluoromethyl)sulphonyl]aniline 97%
3-[(Trifluoromethyl)sulphonyl]aniline
Formula: C₇H₆F₃NO₂S
Melting Pt: 78…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792413
CAS Number: 426-59-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR