N-Methoxy-N-methyl-3-[(trifluoromethyl)sulphanyl]benzamide
Supplier: Apollo Scientific
|
Warning
|
PC48723-1G
PC48723-5G
APOSPC48723-1GEA
72.5
EUR
APOSPC48723-1G
APOSPC48723-5G
N-Methoxy-N-methyl-3-[(trifluoromethyl)sulphanyl]benzamide
N-Methoxy-N-methyl-3-[(trifluoromethyl)sulphanyl]benzamide
Formula:
C₁₀H₁₀F₃NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD28384422 CAS Number: 329942-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...