5-Bromo-3-(trifluoromethyl)picolinic acid 95%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-3-(trifluoromethyl)-2-pyridinecarboxylic acid

PC48747-1G
APOSPC48747-1GEA 98.6 EUR
APOSPC48747-1G
5-Bromo-3-(trifluoromethyl)picolinic acid 95%
5-Bromo-3-(trifluoromethyl)picolinic acid
Formula: C₇H₃BrF₃NO₂
MW: 270.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185433
CAS Number: 1211580-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR