Methyl 6-bromo-5-fluorobenzothiophene-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
PC48756-1G
PC48756-250MG
APOSPC48756-1GEA
580
EUR
APOSPC48756-1G
APOSPC48756-250MG
Methyl 6-bromo-5-fluorobenzothiophene-2-carboxylate
Methyl 6-bromo-5-fluorobenzothiophene-2-carboxylate
Formula:
C₁₀H₆BrFO₂S Storage Temperature: Ambient |
MDL Number:
MFCD29430786 CAS Number: 1404119-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...