2-Chloro-4-fluoro-5-iodoaniline 97%
Supplier: Apollo Scientific
PC48771-1G
PC48771-5G
PC48771-250MG
APOSPC48771-1GEA
429
EUR
APOSPC48771-1G
APOSPC48771-5G
APOSPC48771-250MG
2-Chloro-4-fluoro-5-iodoaniline 97%
2-Chloro-4-fluoro-5-iodoaniline
Formula:
C₆H₄ClFIN MW: 271,459 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31629890 CAS Number: 2091569-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...