tert-Butyl (R)-(1-fluoro-3-methylbutan-2-yl)carbamate 95%
Supplier: Apollo Scientific
|
Warning
|
PC48797-1G
PC48797-250MG
APOSPC48797-1GEA
611
EUR
APOSPC48797-1G
APOSPC48797-250MG
tert-Butyl (R)-(1-fluoro-3-methylbutan-2-yl)carbamate 95%
tert-Butyl (R)-(1-fluoro-3-methylbutan-2-yl)carbamate
Formula:
C₁₀H₂₀FNO₂ MW: 205,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30378948 CAS Number: 942275-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...