Methyl 5-bromo-3-fluoro-2-(trifluoromethyl)benzoate 95%
Supplier: Apollo Scientific
PC48836-1G
PC48836-250MG
APOSPC48836-1GEA
632
EUR
APOSPC48836-1G
APOSPC48836-250MG
Methyl 5-bromo-3-fluoro-2-(trifluoromethyl)benzoate 95%
Methyl 5-bromo-3-fluoro-2-(trifluoromethyl)benzoate
Formula:
C₉H₅BrF₄O₂ MW: 301,032 g/mol Storage Temperature: Ambient |
CAS Number:
2090828-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...