Methyl 3-fluoro-4-sulphamoylbenzoate
Supplier: Apollo Scientific
|
Warning
|
PC48924-1G
PC48924-250MG
APOSPC48924-1GEA
575
EUR
APOSPC48924-1G
APOSPC48924-250MG
Methyl 3-fluoro-4-sulphamoylbenzoate
Methyl 3-fluoro-4-sulphamoylbenzoate
Formula:
C₈H₈FNO₄S MW: 233,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32070713 CAS Number: 1204574-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...