6-Amino-2-bromo-3-fluorophenol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-4-fluoro-2-hydroxyaniline
PC49005-1G
PC49005-5G
APOSPC49005-1GEA
78.3
EUR
APOSPC49005-1G
APOSPC49005-5G
6-Amino-2-bromo-3-fluorophenol
6-Amino-2-bromo-3-fluorophenol
Formula:
C₆H₅BrFNO MW: 206.012 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18071061 CAS Number: 1257535-00-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...