3-Chloro-5-fluoropyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Carboxy-3-chloro-5-fluoropyridine, 3-Chloro-5-fluoropicolinic acid

PC49027-250MG PC49027-100MG
APOSPC49027-250MGEA 205 EUR
APOSPC49027-250MG APOSPC49027-100MG
3-Chloro-5-fluoropyridine-2-carboxylic acid
3-Chloro-5-fluoropyridine-2-carboxylic acid
Formula: C₆H₃ClFNO₂
MW: 175,545 g/mol
Melting Pt: 142…144 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD13185822
CAS Number: 128073-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR