5-Ethynyl-2-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific
Warning

Synonyms: [6-(Trifluoromethyl)pyridin-3-yl]acetylene, 5-Ethynyl-α,α,α-trifluoro-2-picoline

PC49132-1G PC49132-250MG PC49132-5G
APOSPC49132-1GEA 166 EUR
APOSPC49132-1G APOSPC49132-250MG APOSPC49132-5G
5-Ethynyl-2-(trifluoromethyl)pyridine 95%
5-Ethynyl-2-(trifluoromethyl)pyridine
Formula: C₈H₄F₃N
MW: 171,119 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189725
CAS Number: 1196155-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR