2'-Amino-5'-bromo-2,2,2-trifluoroacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(2-Amino-5-bromophenyl)-2,2,2-trifluoroethan-1-one, 4-Bromo-2-(trifluoroacetyl)aniline, 1-(2-Amino-5-bromophenyl)-2,2,2-trifluoroethanone

PC49164-1G PC49164-250MG
APOSPC49164-1GEA 505 EUR
APOSPC49164-1G APOSPC49164-250MG
2'-Amino-5'-bromo-2,2,2-trifluoroacetophenone
2'-Amino-5'-bromo-2,2,2-trifluoroacetophenone
Formula: C₈H₅BrF₃NO
MW: 268,031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18071073
CAS Number: 1233967-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR