5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-fluoro-6-(methylamino)aniline, 5-Bromo-3-fluoro-N1-methylphenylene-1,2-diamine

PC49259-1G PC49259-250MG
APOSPC49259-1GEA 574 EUR
APOSPC49259-1G APOSPC49259-250MG
5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine
5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine
Formula: C₇H₈BrFN₂
MW: 219,054 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415278
CAS Number: 1396503-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR