3-Nitro-4-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific
Danger

Synonyms: 4-(Bromomethyl)-2-nitro-1-(trifluoromethyl)benzene, 4-(Bromomethyl)-2-nitrobenzotrifluoride

PC49353-1G PC49353-250MG
APOSPC49353-1GEA 220 EUR
APOSPC49353-1G APOSPC49353-250MG
3-Nitro-4-(trifluoromethyl)benzyl bromide 98%
3-Nitro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₃NO₂
MW: 284,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606359
CAS Number: 1227572-51-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR